missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisatin 98.0+%, TCI America™

Catalog No. N05725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05725G 5 g
N057225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05725G Supplier TCI America Supplier No. N05725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-09-6
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005720
InChI Key UNMYHYODJHKLOC-UHFFFAOYSA-N
Synonym 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
PubChem CID 4669250
IUPAC Name 5-nitro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2

Specifications

Melting Point 255°C
Quantity 5 g
Formula Weight 192.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisatin
RTECSNumber NL7970000
TSCA No