missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisatin, 98%

CAS: 611-09-6 | C8H4N2O4 | 192.13 g/mol

$75.03 - $236.55

Chemical Identifiers

CAS 611-09-6
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005720
InChI Key UNMYHYODJHKLOC-UHFFFAOYSA-N
Synonym 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
PubChem CID 4669250
IUPAC Name 5-nitro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2496206
View Documents
Thermo Scientific Chemicals
B2496206
5 g
Each for $75.03
Only null left
 
AAB2496214
View Documents
Thermo Scientific Chemicals
B2496214
25 g
Each for $236.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

611-09-6
192.13
UNMYHYODJHKLOC-UHFFFAOYSA-N
4669250
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
C8H4N2O4
MFCD00005720
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
5-nitro-1H-indole-2,3-dione

Specifications

611-09-6
C8H4N2O4
5 g
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
192.13
192.13
5-Nitroisatin
∼252°C (decomposition)
MFCD00005720
180223
UNMYHYODJHKLOC-UHFFFAOYSA-N
5-nitro-1H-indole-2,3-dione
4669250
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302-H312-H332-H340

EINECSNumber : 210-252-5

RTECSNumber : NL7970000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only