missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzimidazole 98.0+%, TCI America™

Catalog No. N015225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N015225G Supplier TCI America Supplier No. N015225G

Chemical Identifiers

CAS 94-52-0
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00005604
InChI Key XPAZGLFMMUODDK-UHFFFAOYSA-N
Synonym 5-nitrobenzimidazole, 6-nitrobenzimidazole, 5-nitro-1h-benzo d imidazole, 5-nitro-1h-benzimidazole, 1h-benzimidazole, 5-nitro, 1h-benzimidazole, 6-nitro, 6-nitro-benzimidazole, 5 6-nitrobenzimidazole, benzimidazole, 5-nitro, benzimidazole, 6-nitro
PubChem CID 7195
IUPAC Name 6-nitro-1H-1,3-benzodiazole
SMILES [O-][N+](=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point 209°C
Color Yellow
Quantity 25 g
Formula Weight 163.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrobenzimidazole
EINECSNumber (5)-0466
RTECSNumber DD9800000
TSCA Yes