missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzimidazole, 98+%

Catalog No. AAA1052430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1052430 250 g
AAA1052418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1052430 Supplier Thermo Scientific Chemicals Supplier No. A1052430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94-52-0 | C7H5N3O2 | 163.14 g/mol

5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.

Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.

Chemical Identifiers

CAS 94-52-0
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00005604
InChI Key XPAZGLFMMUODDK-UHFFFAOYSA-N
Synonym 5-nitrobenzimidazole, 6-nitrobenzimidazole, 5-nitro-1h-benzo d imidazole, 5-nitro-1h-benzimidazole, 1h-benzimidazole, 5-nitro, 1h-benzimidazole, 6-nitro, 6-nitro-benzimidazole, 5 6-nitrobenzimidazole, benzimidazole, 5-nitro, benzimidazole, 6-nitro
PubChem CID 7195
IUPAC Name 6-nitro-1H-benzimidazole
SMILES [O-][N+](=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point 207°C to 211°C
Quantity 250 g
Beilstein 7926
Solubility Information Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Formula Weight 163.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitrobenzimidazole
Hazard Category H302-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 202-341-2
RTECSNumber DD9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only