missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-pyridinecarboxylic Acid 98.0+%, TCI America™
$154.71 - $462.29
Chemical Identifiers
| CAS | 30651-24-2 |
|---|---|
| Molecular Formula | C6H4N2O4 |
| Molecular Weight (g/mol) | 168.11 |
| MDL Number | MFCD04114182 |
| InChI Key | QKYRCTVBMNXTBT-UHFFFAOYSA-N |
| Synonym | 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid |
| PubChem CID | 520488 |
| IUPAC Name | 5-nitropyridine-2-carboxylic acid |
| SMILES | OC(=O)C1=NC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 30651-24-2 | |
| 168.11 | |
| QKYRCTVBMNXTBT-UHFFFAOYSA-N | |
| 520488 | |
| OC(=O)C1=NC=C(C=C1)[N+]([O-])=O |
| C6H4N2O4 | |
| MFCD04114182 | |
| 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid | |
| 5-nitropyridine-2-carboxylic acid |
Specifications
| 30651-24-2 | |
| White-Yellow | |
| MFCD04114182 | |
| 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid | |
| OC(=O)C1=NC=C(C=C1)[N+]([O-])=O | |
| 168.11 | |
| 168.11 | |
| Crystalline Powder |
| 204°C | |
| C6H4N2O4 | |
| 1 g | |
| QKYRCTVBMNXTBT-UHFFFAOYSA-N | |
| 5-nitropyridine-2-carboxylic acid | |
| 520488 | |
| ≥98.0% (HPLC,T) | |
| 5-Nitro-2-pyridinecarboxylic Acid |
Safety and Handling
TSCA : No