missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furonitrile, 97%

Catalog No. p-7045375
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1048606 5 g
AAL1048614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1048606 Supplier Thermo Scientific Chemicals Supplier No. L1048606

CAS: 59-82-5 | C5H2N2O3 | 138.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-82-5
Molecular Formula C5H2N2O3
Molecular Weight (g/mol) 138.08
MDL Number MFCD00082647
InChI Key DUJNJLFQOODDNJ-UHFFFAOYSA-N
Synonym 5-nitro-2-furonitrile, 5-nitro-2-furancarbonitrile, unii-e63b56v8ya, 2-furancarbonitrile, 5-nitro, 5-nitro-2-cyanofuran, 5-nitro-2-furonitril, 2-nitrofuran-5-carbonitrile, acmc-1an56, 5-nitro-furan-2-carbonitrile, 2-furancarbonitrile,5-nitro
PubChem CID 94881
IUPAC Name 5-nitrofuran-2-carbonitrile
SMILES [O-][N+](=O)C1=CC=C(O1)C#N

Specifications

Melting Point 61°C to 64°C
Quantity 5 g
UN Number UN3439
Beilstein 123933
Formula Weight 138.08
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-furonitrile
Hazard Category H302+H332-H311
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only