missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde 98.0+%, TCI America™

Catalog No. p-7133786
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03875G 5 g
N038725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03875G Supplier TCI America Supplier No. N03875G
Only null left

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5H3NO4
Molecular Weight (g/mol) 141.082
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O

Specifications

Quantity 5 g
UN Number 1325
Formula Weight 141.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Nitro-2-furaldehyde
EINECSNumber (9)-0976
RTECSNumber LT7200000
TSCA No