missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde semicarbazone, 98+%

Catalog No. AAA1859330
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1859330 250 g
AAA1859314 25 g
AAA1859322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1859330 Supplier Thermo Scientific Chemicals Supplier No. A1859330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-87-0 | C6H6N4O4 | 198.14 g/mol

Bactericidal compound used as an antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-87-0
Molecular Formula C6H6N4O4
Molecular Weight (g/mol) 198.14
MDL Number MFCD00003225
InChI Key IAIWVQXQOWNYOU-FPYGCLRLSA-N
Synonym nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n
PubChem CID 5447130
IUPAC Name [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
SMILES NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 250 g
Beilstein 86403
Sensitivity Light sensitive
Merck Index 14,6600
Formula Weight 198.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-2-furaldehyde semicarbazone
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H341-H351-H361
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 200-443-1
RTECSNumber LT7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only