missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde Diacetate 97.0+%, TCI America™

Catalog No. N021825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N021825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N021825G Supplier TCI America Supplier No. N021825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-55-7
Molecular Formula C9H9NO7
Molecular Weight (g/mol) 243.171
MDL Number MFCD00003244
InChI Key HSXKWKJCZNRMJO-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate
PubChem CID 7097
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
SMILES CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 25 g
Formula Weight 243.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitro-2-furaldehyde Diacetate
EINECSNumber (5)-0045
RTECSNumber LU1940000
TSCA Yes