missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-furaldehyde diacetate, 98%
CAS: 92-55-7 | C9H9NO7 | 243.17 g/mol
$63.62 - $63.62
Chemical Identifiers
| CAS | 92-55-7 |
|---|---|
| Molecular Formula | C9H9NO7 |
| Molecular Weight (g/mol) | 243.17 |
| MDL Number | MFCD00003244 |
| InChI Key | HSXKWKJCZNRMJO-UHFFFAOYSA-N |
| Synonym | 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate |
| PubChem CID | 7097 |
| IUPAC Name | [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate |
| SMILES | CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC165131000
|
Thermo Scientific Chemicals
165131000 |
100 g | Glass bottle |
Each for $63.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 92-55-7 | |
| 243.17 | |
| HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
| 7097 | |
| CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C |
| C9H9NO7 | |
| MFCD00003244 | |
| 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
| [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate |
Specifications
| 92-55-7 | |
| 100.0 | |
| Yellow | |
| 97.5% min. (GC) | |
| C9H9NO7 | |
| 100 g | |
| HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
| [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate | |
| 7097 | |
| 98% | |
| 5-Nitro-2-furaldehyde diacetate, 98% |
| 97.5 | |
| 89.0°C to 93.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00003244 | |
| 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
| CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C | |
| 243.17 | |
| 243.17 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 202-166-1
RTECSNumber : LU1940000
TSCA : TSCA
RUO – Research Use Only