missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde diacetate, 98%

Catalog No. AC165135000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165135000 500 g Glass bottle
AC165131000 100 g Glass bottle
AC165130025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165135000 Supplier Thermo Scientific Chemicals Supplier No. 165135000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-55-7 | C9H9NO7 | 243.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-55-7
Molecular Formula C9H9NO7
Molecular Weight (g/mol) 243.17
MDL Number MFCD00003244
InChI Key HSXKWKJCZNRMJO-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate
PubChem CID 7097
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
SMILES CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 500 g
Formula Weight 243.17
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Nitro-2-furaldehyde diacetate, 98%
Show More Show Less
EINECSNumber 202-166-1
RTECSNumber LU1940000
TSCA TSCA

RUO – Research Use Only