missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde diacetate, 97%

Catalog No. p-7024023
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1816922 100 g
AAA1816936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1816922 Supplier Thermo Scientific Chemicals Supplier No. A1816922

CAS: 92-55-7 | C9H9NO7 | 243.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-55-7
Molecular Formula C9H9NO7
Molecular Weight (g/mol) 243.171
MDL Number MFCD00003244
InChI Key HSXKWKJCZNRMJO-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate
PubChem CID 7097
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
SMILES CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C

Specifications

Melting Point 87°C to 92°C
Quantity 100 g
Beilstein 255089
Sensitivity Light sensitive
Formula Weight 243.17
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-2-furaldehyde diacetate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 202-166-1
RTECSNumber LU1940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only