missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1,10-phenanthroline, 98%

Catalog No. p-7081444
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1142803 1 g
AAA1142806 5 g
AAA1142814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1142803 Supplier Thermo Scientific Chemicals Supplier No. A1142803
Only null left

CAS: 4199-88-6 | C12H7N3O2 | 225.207 g/mol

5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4199-88-6
Molecular Formula C12H7N3O2
Molecular Weight (g/mol) 225.207
MDL Number MFCD00004981
InChI Key PDDBTWXLNJNICS-UHFFFAOYSA-N
PubChem CID 72790
IUPAC Name 5-nitro-1,10-phenanthroline
SMILES C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]

Specifications

Melting Point 202°C to 204°C
Quantity 1 g
Beilstein 196245
Synonym 1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 225.21
Percent Purity 98%
Chemical Name or Material 5-Nitro-1,10-phenanthroline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 224-097-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only