missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1,10-phenanthroline, 98%

Catalog No. p-3779318
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC186080010 1 g Glass bottle
1 options

Catalog No. AC186080010

Supplier: Thermo Scientific Chemicals 186080010

Only null left

CAS: 4199-88-6 | C12H7N3O2 | 225.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4199-88-6
Molecular Formula C12H7N3O2
Molecular Weight (g/mol) 225.21
MDL Number MFCD00004981
InChI Key PDDBTWXLNJNICS-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
PubChem CID 72790
IUPAC Name 5-nitro-1,10-phenanthroline
SMILES C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 199.0°C to 203.0°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 225.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitro-1, 10-phenanthroline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-097-6

RUO – Research Use Only