Chemglass Inc Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange
Catalog No. CG194604
Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.