missing translation for 'onlineSavingsMsg'
Learn More

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, MP Biomedicals™

Catalog No. MP215775025
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215775025 25 g
MP215775080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215775025 Supplier MP Biomedicals Inc Supplier No. 0215775025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, White Powder

  • It is useful as a nonionic density gradient medium.
  • It is a Non-ionic monomeric triiodinated x-ray contrast agent

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9
Molecular Weight (g/mol) 821.14
MDL Number MFCD00077732
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
Synonym Nycodenz, Histodenz™

Specifications

Color White
Quantity 25 g
Beilstein 2406632
Physical Form Powder
Assay 0.99
RTECSNumber CZ2205000
storageNote1 Store at -0°C.