missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. p-3881851
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC73404CB 250mg Amber Glass Bottle
CC73404DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC73404CB Supplier Thermo Scientific Chemicals Supplier No. CC73404CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 893744-01-9
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD06803315
InChI Key YPXGCMYNDMFOHE-UHFFFAOYSA-N
Synonym 2-formyl-5-morpholin-4-ylmethyl thiophene, 5-morpholinomethyl thiophene-2-carbaldehyde, 5-morpholin-4-ylmethyl thiophene-2-carbaldehyde, 5-4-morpholinylmethyl-2-thiophenecarbaldehyde, 2-thiophenecarboxaldehyde,5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl thiophene-2-carboxaldehyde, 5-morpholin-4-yl methyl thiophene-2-carbaldehyde
PubChem CID 20098933
IUPAC Name 5-(morpholin-4-ylmethyl)thiophene-2-carbaldehyde
SMILES O=CC1=CC=C(CN2CCOCC2)S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 211.28
Percent Purity 97%
Chemical Name or Material 5-(Morpholinomethyl)thiophene-2-carbaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.