missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)isoxazole-3-carbaldehyde, Thermo Scientific™

Catalog No. p-4391433
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70804CB 250mg Amber Glass Bottle
CC70804DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC70804CB Supplier Thermo Scientific Chemicals Supplier No. CC70804CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 893749-85-4
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.206
InChI Key AGLGFPIEVWZSLU-UHFFFAOYSA-N
Synonym 5-morpholinomethyl isoxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl-1,2-oxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl isoxazole-3-carbaldehyde, 5-morpholin-4-yl methyl-1,2-oxazole-3-carbaldehyde
PubChem CID 20099059
IUPAC Name 5-(morpholin-4-ylmethyl)-1,2-oxazole-3-carbaldehyde
SMILES C1COCCN1CC2=CC(=NO2)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 5-(morpholinomethyl)isoxazole-3-carbaldehyde
Hazard Category GHS Signal Word: Danger