missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(morpholinomethyl)isoxazole-3-carbaldehyde, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 196.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-(morpholinomethyl)isoxazole-3-carbaldehyde |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |