missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)-2-furaldehyde, 97%, Thermo Scientific™

Catalog No. p-3881848
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC72504CB 250mg Amber Glass Bottle
CC72504DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC72504CB Supplier Thermo Scientific Chemicals Supplier No. CC72504CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 392659-97-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD01821235
InChI Key ZQILSGYYJOBENS-UHFFFAOYSA-N
Synonym 5-morpholinomethyl-2-furaldehyde, 5-morpholin-4-ylmethyl furan-2-carbaldehyde, 5-morpholin-4-ylmethyl-furan-2-carbaldehyde, 5-morpholin-4-ylmethyl furan-2-carboxaldehyde, 2-furancarboxaldehyde,5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl-2-furaldehyde, 5-morpholinomethyl furan-2-carbaldehyde, 5-morpholin-4-yl methyl furan-2-carbaldehyde
PubChem CID 3159700
IUPAC Name 5-(morpholin-4-ylmethyl)furan-2-carbaldehyde
SMILES C1COCCN1CC2=CC=C(O2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material 5-(Morpholinomethyl)-2-furaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.