Learn More
Greiner Bio-One 5 mL Sterile Serological Pipets
Drip-free pipetting design with high optical clarity and maximum accuracy.
Supplier: Greiner Bio-One 606180
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
| Polystyrene | |
| Yes | |
| Individual Paper/Plastic Wrap | |
| Serological Pipet |
| Sterile | |
| 1/10 | |
| 5 mL |