missing translation for 'onlineSavingsMsg'
Learn More

5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™

Catalog No. AC158670010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC158670010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC158670010 Supplier Thermo Scientific Chemicals Supplier No. 158670010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

Molecular Formula C11H15N2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00012683
InChI Key PYOUAIQXJALPKW-UHFFFAOYSA-O
Synonym 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
PubChem CID 6917145
SMILES CC1=CC=C2NC=C(CC[NH3+])C2=C1

Specifications

CAS 1010-95-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 281°C to 283°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride
Formula Weight 210.71
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyltryptamine,hydrochloride
Show More Show Less
EINECSNumber 213-777-8

RUO – Research Use Only