missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carboxaldehyde 97.0+%, TCI America™

Catalog No. M09115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09115G 5 g
M091125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09115G Supplier TCI America Supplier No. M09115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13679-70-4
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005434
InChI Key VAUMDUIUEPIGHM-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde
PubChem CID 61663
IUPAC Name 5-methylthiophene-2-carbaldehyde
SMILES CC1=CC=C(S1)C=O

Specifications

Color Brown-Yellow
Boiling Point 205°C
Quantity 5 g
Formula Weight 126.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde