missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6110603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6110603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6110603 Supplier Thermo Scientific Chemicals Supplier No. H6110603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20873-58-9 | Molecular Formula: C6H6O2S2 | Molecular Weight: 174.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20873-58-9
Molecular Formula C6H6O2S2
Molecular Weight (g/mol) 174.232
MDL Number MFCD00173742
InChI Key FWFWUSLBIIIIEN-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carboxylic acid, 5-methylsulfanyl thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-methylthio, 2-thiophenecarboxylicacid, 5-methylthio, 5-methylthiothiophene-2-carboxylic acid, maybridge1_007717, d04ngc, cambridge id 5103856, 5-methylmercaptothiophene-2-carboxylic, 5-methylthio-2-thiophenecarboxylic acid
PubChem CID 714372
IUPAC Name 5-methylsulfanylthiophene-2-carboxylic acid
SMILES CSC1=CC=C(S1)C(=O)O

Specifications

Melting Point 103°C to 105°C
Color White
Quantity 1 g
Formula Weight 174.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Methylthio)thiophene-2-carboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only