missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazole 98.0+%, TCI America™

Catalog No. p-7133104
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22345G 5 g
M223425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22345G Supplier TCI America Supplier No. M22345G
Only null left

Chemical Identifiers

CAS 3581-89-3
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00005335
InChI Key RLYUNPNLXMSXAX-UHFFFAOYSA-N
Synonym 5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
PubChem CID 137980
IUPAC Name 5-methyl-1,3-thiazole
SMILES CC1=CN=CS1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 1993
Formula Weight 99.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methylthiazole