missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazole, 97%

Catalog No. p-7046701
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1587906 5 g
AAL1587914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1587906 Supplier Thermo Scientific Chemicals Supplier No. L1587906
Only null left

CAS: 3581-89-3 | C4H5NS | 99.151 g/mol

5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3581-89-3
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00005335
InChI Key RLYUNPNLXMSXAX-UHFFFAOYSA-N
Synonym 5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
PubChem CID 137980
IUPAC Name 5-methyl-1,3-thiazole
SMILES CC1=CN=CS1

Specifications

Density 1.12
Boiling Point 141°C to 142°C
Flash Point 42°C (107°F)
Odor Stench
Refractive Index 1.528
Quantity 5 g
UN Number UN1993
Beilstein 104674
Solubility Information Insoluble in water.
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material 5-Methylthiazole
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only