missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7132801
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16765G 5 g
M167625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16765G Supplier TCI America Supplier No. M16765G

Chemical Identifiers

CAS 613-84-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00274297
InChI Key ILEIUTCVWLYZOM-UHFFFAOYSA-N
Synonym 5-methylsalicylaldehyde, benzaldehyde, 2-hydroxy-5-methyl, 2,5-cresotaldehyde, 5-methylsalicylic aldehyde, 2-hydroxy-5-methyl-benzaldehyde, 5methylsalicylaldehyde, pubchem15680, 5-methyl salicylaldehyde, 4-hydroxy-m-tolualdehyde, acmc-1bh2k
PubChem CID 69184
IUPAC Name 2-hydroxy-5-methylbenzaldehyde
SMILES CC1=CC=C(O)C(C=O)=C1

Specifications

Melting Point 56°C
Boiling Point 218°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylsalicylaldehyde