missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-3-boronic acid, 98%

Catalog No. p-7035177
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52891MD 250 mg
AAH5289103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52891MD Supplier Thermo Scientific Chemicals Supplier No. H52891MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 173999-18-3 | C6H8BNO2 | 136.945 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 173999-18-3
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD03428528
InChI Key REONQWGHSQHTAC-UHFFFAOYSA-N
Synonym 5-methylpyridine-3-boronic acid, 5-methyl-3-pyridineboronic acid, 5-methylpyridin-3-yl boronic acid, 5-methylpyridin-3-yl-3-boronic acid, 5-methylpyridine-3-boronicacid, 5-methyl-3-pyridyl boronic acid, 5-methyl-3-pyridinyl boronic acid, pubchem17082, 3-borono-5-methylpyridine, acmc-209e7n
PubChem CID 2762708
IUPAC Name (5-methylpyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1)C)(O)O

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 136.95
Percent Purity 98%
Chemical Name or Material 5-Methylpyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only