missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinic acid, 97%

CAS: 3222-49-9 | C7H7NO2 | 137.138 g/mol

Supplier:  Thermo Scientific Chemicals L1637006

 View more versions of this product

Catalog No. AAL1637006


Only null left
Explore available promotions

Description

Description

5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Sparingly soluble (0.083 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

5-Methylnicotinic acid
210°C to 215°C
MFCD00829036
112288
Sparingly soluble (0.083g/L at 25°C).
CC1=CN=CC(=C1)C(=O)O
137.138
137.14
3222-49-9
C7H7NO2
5 g
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
DJDHHXDFKSLEQY-UHFFFAOYSA-N
5-methylpyridine-3-carboxylic acid
256208
97%

Chemical Identifiers

3222-49-9
137.138
DJDHHXDFKSLEQY-UHFFFAOYSA-N
256208
CC1=CN=CC(=C1)C(=O)O
C7H7NO2
MFCD00829036
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
5-methylpyridine-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only