missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinic acid, 97%

Catalog No. AAL1637003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1637003 1 g
AAL1637006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1637003 Supplier Thermo Scientific Chemicals Supplier No. L1637003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3222-49-9 | C7H7NO2 | 137.138 g/mol

5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Sparingly soluble (0.083 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3222-49-9
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00829036
InChI Key DJDHHXDFKSLEQY-UHFFFAOYSA-N
Synonym 5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
PubChem CID 256208
IUPAC Name 5-methylpyridine-3-carboxylic acid
SMILES CC1=CN=CC(=C1)C(=O)O

Specifications

Melting Point 210°C to 215°C
Quantity 1 g
Beilstein 112288
Solubility Information Sparingly soluble (0.083g/L at 25°C).
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only