missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-3-carbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH5069303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5069303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5069303 Supplier Thermo Scientific Chemicals Supplier No. H5069303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 39499-34-8 | Molecular Formula: C5H4ClNO2 | Molecular Weight: 145.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39499-34-8
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.54
MDL Number MFCD00085127
InChI Key XMVNMWDLOGSUSM-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carbonyl chloride, 3-isoxazolecarbonyl chloride, 5-methyl, 5-methyl-3-isoxazolecarbonyl chloride, 5-methyl-isoxazole-3-carbonyl chloride, acmc-209j5v, 5-methyl-3-isoxazoyl chloride, 5-methylisoxazole carbonyl chloride, 5-methylisoxazol-3-ylcarbonyl chloride, 3-isoxazolecarbonylchloride, 5-methyl, 5-methyl isoxazole-3-carbonyl chloride
PubChem CID 2736894
IUPAC Name 5-methyl-1,2-oxazole-3-carbonyl chloride
SMILES CC1=CC(=NO1)C(Cl)=O

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 145.55
Percent Purity 98%
Chemical Name or Material 5-Methylisoxazole-3-carbonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-475-6
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only