missing translation for 'onlineSavingsMsg'
Learn More

(5-Methylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC10409DE

Quantity 5g

 View more versions of this product

Catalog No. CC10409DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

35166-33-7
113.12
MDYHWQQHEWDJKR-UHFFFAOYSA-N
2736567
CC1=CC(CO)=NO1
C5H7NO2
MFCD00085129
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
(5-methyl-1,2-oxazol-3-yl)methanol

Specifications

(5-Methylisoxazol-3-yl)methanol
96.5
97%
5g
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
CC1=CC(CO)=NO1
113.12
113.12
35166-33-7
100.0
C5H7NO2
MFCD00085129
MDYHWQQHEWDJKR-UHFFFAOYSA-N
(5-methyl-1,2-oxazol-3-yl)methanol
2736567
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents