missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin 99.0+%, TCI America™

Supplier:  TCI America M170325G

 View more versions of this product

Catalog No. M170325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Methylisatin
188°C
MFCD00005721
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
CC1=CC=C2NC(=O)C(=O)C2=C1
161.16
161.16
Crystalline Powder
608-05-9
C9H7NO2
25 g
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methyl-2,3-dihydro-1H-indole-2,3-dione
11840
≥99.0% (GC)

Chemical Identifiers

608-05-9
161.16
VAJCSPZKMVQIAP-UHFFFAOYSA-N
11840
CC1=CC=C2NC(=O)C(=O)C2=C1
C9H7NO2
MFCD00005721
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
5-methyl-2,3-dihydro-1H-indole-2,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3387

RTECSNumber : NL7939300

TSCA : No

SDS
Documents

Documents

Promotions

Promotions