missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin, 97%

CAS: 608-05-9 | C9H7NO2 | 161.16 g/mol

Supplier:  Thermo Scientific Chemicals L0800914

 View more versions of this product

Catalog No. AAL0800914


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Methylisatin
∽180°C (decomposition)
MFCD00005721
123738
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methyl-1H-indole-2,3-dione
11840
97%
608-05-9
C9H7NO2
25 g
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
CC1=CC=C2NC(=O)C(=O)C2=C1
161.16
161.16

Chemical Identifiers

608-05-9
161.16
VAJCSPZKMVQIAP-UHFFFAOYSA-N
11840
CC1=CC=C2NC(=O)C(=O)C2=C1
C9H7NO2
MFCD00005721
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
5-methyl-1H-indole-2,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-152-1

RTECSNumber : NL7939300

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only