missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin, 97%

Catalog No. AAL0800903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0800903 1 g
AAL0800906 5 g
AAL0800914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0800903 Supplier Thermo Scientific Chemicals Supplier No. L0800903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608-05-9 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-05-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005721
InChI Key VAJCSPZKMVQIAP-UHFFFAOYSA-N
Synonym 5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
PubChem CID 11840
IUPAC Name 5-methyl-1H-indole-2,3-dione
SMILES CC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Beilstein 123738
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 5-Methylisatin
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-152-1
RTECSNumber NL7939300
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only