missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindoline 98.0+%, TCI America™

Catalog No. M13725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13725G Supplier TCI America Supplier No. M13725G
Only null left

Chemical Identifiers

CAS 65826-95-1
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD03093636
InChI Key JFUAVVHABJWSFX-UHFFFAOYSA-N
PubChem CID 14023679
IUPAC Name 5-methyl-2,3-dihydro-1H-indole
SMILES CC1=CC2=C(C=C1)NCC2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Methylindoline