missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole-2-carboxylic acid, 99%

Catalog No. AAL1714503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1714503 1 g
AAL1714506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1714503 Supplier Thermo Scientific Chemicals Supplier No. L1714503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10241-97-1 | C10H9NO2 | 175.19 g/mol

5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists

Solubility
Soluble in ethanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 10241-97-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00047166
InChI Key DAITVOCMWPNFTL-UHFFFAOYSA-N
Synonym 5-methylindole-2-carboxylic acid, 5-methyl-2-indolecarboxylic acid, chembl23328, 1h-indole-2-carboxylic acid, 5-methyl, acmc-1bxae, 5-me-ica, daitvocmwpnftl-uhfffaoysa, 5-methyl indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 5-methyl, #
PubChem CID 259188
IUPAC Name 5-methyl-1H-indole-2-carboxylic acid
SMILES CC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

Melting Point ∽235°C (decomposition)
Quantity 1 g
Beilstein 144055
Solubility Information Soluble in ethanol (50mg/ml). Insoluble in water.
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material 5-Methylindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only