missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfurylamine 98.0+%, TCI America™

Catalog No. M13115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13115G 5 g
M131125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13115G Supplier TCI America Supplier No. M13115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14003-16-8
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00143471
InChI Key YSEAGSCGERFGBL-UHFFFAOYSA-N
Synonym 5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz
PubChem CID 2724683
IUPAC Name 1-(5-methylfuran-2-yl)methanamine
SMILES CC1=CC=C(CN)O1

Specifications

Color Yellow
Boiling Point 55°C
Quantity 5 g
UN Number 2733
Formula Weight 111.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Methylfurfurylamine