missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylfurfural, 98+%
CAS: 620-02-0 | C6H6O2 | 110.11 g/mol
$97.68 - $97.68
Chemical Identifiers
| CAS | 620-02-0 |
|---|---|
| Molecular Formula | C6H6O2 |
| Molecular Weight (g/mol) | 110.11 |
| MDL Number | MFCD00003232 |
| InChI Key | OUDFNZMQXZILJD-UHFFFAOYSA-N |
| Synonym | 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran |
| PubChem CID | 12097 |
| ChEBI | CHEBI:2091 |
| IUPAC Name | 5-methylfuran-2-carbaldehyde |
| SMILES | CC1=CC=C(O1)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC159920250
|
Thermo Scientific Chemicals
159920250 |
25 g | Glass bottle |
Each for $97.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 620-02-0 | |
| 110.11 | |
| OUDFNZMQXZILJD-UHFFFAOYSA-N | |
| 12097 | |
| 5-methylfuran-2-carbaldehyde |
| C6H6O2 | |
| MFCD00003232 | |
| 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
| CHEBI:2091 | |
| CC1=CC=C(O1)C=O |
Specifications
| 620-02-0 | |
| 98.0 | |
| Yellow to Brown | |
| 187°C | |
| Authentic | |
| Glass bottle | |
| 1.5290 to 1.532 | |
| 25 g | |
| 1.1 | |
| Solubility in water: 33.3g/L (20 c) | |
| CC1=CC=C(O1)C=O | |
| 110.11 | |
| CHEBI:2091 | |
| 98+% | |
| 5-Methylfurfural, 98+% |
| 10mg KOH/g max. | |
| 100.0 | |
| 1.1000g/mL | |
| 72°C | |
| 98% min. (GC) | |
| C6H6O2 | |
| MFCD00003232 | |
| 17, 289 | |
| 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
| OUDFNZMQXZILJD-UHFFFAOYSA-N | |
| 5-methylfuran-2-carbaldehyde | |
| 12097 | |
| 110.11 | |
| Liquid |
Safety and Handling
EINECSNumber : 210-622-6
RTECSNumber : LT7032500
TSCA : TSCA
RUO – Research Use Only