missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, 98+%

Catalog No. AC159921000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159921000 100 g Glass bottle
AC159920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159921000 Supplier Thermo Scientific Chemicals Supplier No. 159921000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 620-02-0 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Acidity 10mg KOH/g max.
CAS Min % 98.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1000g/mL
Boiling Point 187°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 100 g
Beilstein 17, 289
Specific Gravity 1.1
Solubility Information Solubility in water: 33.3g/L (20 c)
Formula Weight 110.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 5-Methylfurfural, 98+%
Show More Show Less
EINECSNumber 210-622-6
RTECSNumber LT7032500
TSCA TSCA

RUO – Research Use Only