missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcytidine 98.0+%, TCI America™

Catalog No. M19311G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19311G Supplier TCI America Supplier No. M19311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2140-61-6
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.25
MDL Number MFCD00006548
InChI Key ZAYHVCMSTBRABG-JXOAFFINSA-N
PubChem CID 92918
ChEBI CHEBI:20607
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2-dihydropyrimidin-2-one
SMILES CC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N=C1N

Specifications

Melting Point 211°C
Color White-Yellow
Quantity 1 g
Formula Weight 257.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylcytidine