missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-DL-tryptophan 98.0+%, TCI America™

Catalog No. p-7132180
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0452100MG 100 mg
M04521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0452100MG Supplier TCI America Supplier No. M0452100MG
Only null left

Chemical Identifiers

CAS 951-55-3
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.26
MDL Number MFCD00005652
InChI Key HUNCSWANZMJLPM-UHFFFAOYSA-N
Synonym 5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652
PubChem CID 92852
ChEBI CHEBI:52524
IUPAC Name 2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
SMILES CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1

Specifications

Melting Point 275°C
Quantity 100 mg
Formula Weight 218.26
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-DL-tryptophan
RTECSNumber YN7140000
TSCA No