missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-5-phenylhydantoin, 99%, Thermo Scientific™

Catalog No. p-3758320
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177680050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC177680050 Supplier Thermo Scientific Chemicals Supplier No. 177680050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6843-49-8
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.2
MDL Number MFCD00005265
InChI Key JNGWGQUYLVSFND-UHFFFAOYSA-N
Synonym 5-methyl-5-phenylhydantoin, hydantoin, 5-methyl-5-phenyl, 2,4-imidazolidinedione,5-methyl-5-phenyl, 2,4-imidazolidinedione, 5-methyl-5-phenyl, mpid standard, acmc-20m4pt, 2,4-imidazolidinedione, 5-methyl-5-phenyl-, r, 2, 5-methyl-5-phenyl, +/--5-methyl-5-phenylhydantoin
PubChem CID 93043
IUPAC Name 5-methyl-5-phenylimidazolidine-2,4-dione
SMILES CC1(C(=O)NC(=O)N1)C2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5g
Beilstein 24, I, 347
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-5-phenylhydantoin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 229-928-6

RUO – Research Use Only