missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride 97.0+%, TCI America™

Catalog No. p-7133451
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2819200MG 200 mg
M28191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2819200MG Supplier TCI America Supplier No. M2819200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 720720-96-7
Molecular Formula C8H11ClN2O2S
Molecular Weight (g/mol) 234.698
MDL Number MFCD25542379
InChI Key ZWIYEBIMFPQYDI-UHFFFAOYSA-N
PubChem CID 11149031
IUPAC Name 5-methyl-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylic acid;hydrochloride
SMILES CN1CCC2=C(C1)SC(=N2)C(=O)O.Cl

Specifications

Melting Point 201°C
Color Yellow
Quantity 200 mg
Formula Weight 234.70
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride