missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carbonyl chloride, 90%, Thermo Scientific™

Catalog No. CC00502DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00502DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00502DA Supplier Thermo Scientific Chemicals Supplier No. CC00502DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16883-16-2
Molecular Formula C11H8ClNO2
Molecular Weight (g/mol) 221.64
MDL Number MFCD00052205
InChI Key HXEVQMXCHCDPSO-UHFFFAOYSA-N
Synonym 5-methyl-3-phenylisoxazole-4-carbonyl chloride, pmic chloride, unii-97ot2t1w6v, 3-phenyl-5-methylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarbonyl chloride, 3-phenyl-5-methyl-isoxazole-4-carbonyl, pubchem8723, acmc-20am25, dsstox_cid_31452
PubChem CID 85624
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Density 1.2700g/mL
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.64
Percent Purity 90%
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.