missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-3-isoxazolyl)methylamine, 97+%, Thermo Scientific™

Catalog No. p-4510021
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10413DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC10413DA Supplier Thermo Scientific Chemicals Supplier No. CC10413DA

Chemical Identifiers

CAS 154016-48-5
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
InChI Key AZVWIMLQRLKLHH-UHFFFAOYSA-N
PubChem CID 2776306
IUPAC Name (5-methyl-1,2-oxazol-3-yl)methanamine
SMILES CC1=CC(=NO1)CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-methyl-3-isoxazolyl methylamine, 5-methylisoxazol-3-yl methanamine, 5-methyl-1,2-oxazol-3-yl methanamine, 5-methylisoxazol-3-yl methylamine, 1-5-methyl-1,2-oxazol-3-yl methanamine, 1-5-methylisoxazol-3-yl methanamine, 3-isoxazolemethanamine,5-methyl, 3-isoxazolemethanamine, 5-methyl, c-5-methyl-isoxazol-3-yl-methylamine, acmc-20a4az
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (5-Methyl-3-isoxazolyl)methylamine