missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-isoxazolecarbonitrile, 97%, Thermo Scientific™

Catalog No. CC10416DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10416DA 1g Amber Glass Bottle
CC10416DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10416DA Supplier Thermo Scientific Chemicals Supplier No. CC10416DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57351-99-2
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.10
MDL Number MFCD09064940
InChI Key JTPIMYCMWLOEDD-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carbonitrile, 5-methyl-3-isoxazolecarbonitrile, 5-methyl-isoxazole-3-carbonitrile, 3-cyano-5-methylisoxazole, 3-isoxazolecarbonitrile,5-methyl, 3-isoxazolecarbonitrile, 5-methyl, 5-?methylisoxazole-?3-?carbonitrile
PubChem CID 24229456
IUPAC Name 5-methyl-1,2-oxazole-3-carbonitrile
SMILES CC1=CC(=NO1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 108.1
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-isoxazolecarbonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.