missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 179.14 |
| Percent Purity | 97% |
| Chemical Name or Material | [5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine |