missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00701DE
| Quantity | 5g |
|---|
Chemical Identifiers
| 17515-74-1 | |
| 194.11 | |
| NURNFMNHTHXTDS-UHFFFAOYSA-N | |
| 2782183 | |
| CC1=CC(C(O)=O)=C(O1)C(F)(F)F |
| C7H5F3O3 | |
| MFCD00275563 | |
| 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl | |
| 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid |
Specifications
| 5-Methyl-2-(trifluoromethyl)-3-furoic acid | |
| 97% | |
| MFCD00275563 | |
| 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl | |
| CC1=CC(C(O)=O)=C(O1)C(F)(F)F | |
| 194.11 | |
| 194.11 |
| 17515-74-1 | |
| C7H5F3O3 | |
| 5g | |
| NURNFMNHTHXTDS-UHFFFAOYSA-N | |
| 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid | |
| 2782183 | |
| 97% |