missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC00703DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00703DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00703DA Supplier Thermo Scientific Chemicals Supplier No. CC00703DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 306935-02-4
Molecular Formula C6H4ClF3O3S
Molecular Weight (g/mol) 248.6
InChI Key GSYXWDYWBRICSD-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl furan-3-sulfonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonyl chloride, 3-furansulfonylchloride, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl furan-3-sulphonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonylchloride, 5-methyl-2-trifluoromethyl-furan-3-sulfonyl chloride, chloro 5-methyl-2-trifluoromethyl 3-furyl sulfone, 3-furansulfonyl chloride, 5-methyl-2-trifluoromethyl-9ci
PubChem CID 2779895
IUPAC Name 5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
SMILES CC1=CC(=C(O1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride